Assay Details
| Gene Symbol: TTLL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005551436.2 | XP_005551493.1 | 2 - 3 | 576 | 78 | |
| GenBank mRNA | AB169207.1 | - | 2 - 3 | 267 | 78 | |
More Information
Set Membership: |
|

TTLL2
Genomic Map