Assay Details
Gene Symbol: TENT5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005552485.2 | XP_005552542.1 | 2 - 3 | 1097 | 98 | |
XM_005552486.2 | XP_005552543.1 | 2 - 3 | 876 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
