Assay Details
| Gene Symbol: ADA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005568054.2 | XP_005568111.1 | 2 - 3 | 598 | 94 | |
| XM_005568055.2 | XP_005568112.1 | 2 - 3 | 342 | 94 | ||
More Information
Set Membership: |
|

ADA2
Genomic Map