Assay Details
| Gene Symbol: KRCC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005575354.2 | XP_005575411.1 | 3 - 4 | 289 | 87 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005575354.2 transcripts.
Set Membership: |
|

KRCC1
Genomic Map