Assay Details
Gene Symbol: MS4A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005577704.2 | XP_005577761.1 | 7 - 8 | 1125 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
