Assay Details
| Gene Symbol: LOC102131932 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005581333.2 | XP_005581390.1 | 1 | 418 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102131932| Gene Symbol: LOC102131932 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005581333.2 | XP_005581390.1 | 1 | 418 | 77 | |
Set Membership: |
|