Assay Details
| Gene Symbol: ATP5PD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005584899.2 | XP_005584956.1 | 1 - 2 | 321 | 81 | |
| XM_005584900.2 | XP_005584957.1 | 1 - 2 | 320 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005584899.2, XM_005584900.2 transcripts.
Set Membership: |
|

ATP5PD
Genomic Map