Assay Details
| Gene Symbol: LOC102133268 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005591430.1 | XP_005591487.1 | 10 - 11 | 1553 | 62 | |
| XM_005591431.2 | XP_005591488.1 | 10 - 11 | 1962 | 62 | ||
More Information
Set Membership: |
|

LOC102133268
Genomic Map