Assay Details
Gene Symbol: LOC102133268 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005591430.1 | XP_005591487.1 | 24 - 25 | 4077 | 126 | |
XM_005591431.2 | XP_005591488.1 | 24 - 25 | 4486 | 126 | ||
GenBank mRNA | AB070190.1 | - | 7 - 8 | 895 | 126 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
