Assay Details
| Gene Symbol: LOC102133268 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005591430.1 | XP_005591487.1 | 30 - 31 | 5014 | 85 | |
| XM_005591431.2 | XP_005591488.1 | 30 - 31 | 5423 | 85 | ||
| GenBank mRNA | AB070033.1 | - | 11 - 12 | 1856 | 85 | |
| AB070190.1 | - | 13 - 14 | 1832 | 85 | ||
More Information
Set Membership: |
|

LOC102133268
Genomic Map