Assay Details
| Gene Symbol: RHOXF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005594475.2 | XP_005594532.1 | 2 - 3 | 355 | 82 | |
| XM_005594476.2 | XP_005594533.1 | 2 - 3 | 355 | 82 | ||
More Information
Set Membership: |
|

RHOXF2
Genomic Map