Assay Details
| Gene Symbol: ZNF496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015430080.1 | XP_015285566.1 | 5 - 6 | 974 | 82 | |
| XM_015430146.1 | XP_015285632.1 | 5 - 6 | 927 | 82 | ||
More Information
Set Membership: |
|

ZNF496
Genomic Map