Assay Details
| Gene Symbol: ERICH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005543006.2 | XP_005543063.1 | 11 - 12 | 1928 | 73 | |
| XM_005543007.2 | XP_005543064.1 | 11 - 12 | 1927 | 73 | ||
| XM_005543008.2 | XP_005543065.1 | 6 - 7 | 1173 | 73 | ||
| XM_015434030.1 | XP_015289516.1 | 7 - 8 | 1356 | 73 | ||
| XM_015434032.1 | XP_015289518.1 | 6 - 7 | 1334 | 73 | ||
More Information
Set Membership: |
|

ERICH3
Genomic Map