Assay Details
| Gene Symbol: ATAD3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015443960.1 | XP_015299446.1 | 4 - 5 | 580 | 62 | |
| XM_015443962.1 | XP_015299448.1 | 4 - 5 | 580 | 62 | ||
More Information
Set Membership: |
|

ATAD3A
Genomic Map