Assay Details
Gene Symbol: TSGA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001319628.1 | NP_001306557.1 | 4 - 5 | 588 | 67 | |
XM_005550781.2 | XP_005550838.1 | 4 - 5 | 626 | 67 | ||
XM_005550782.2 | XP_005550839.1 | 4 - 5 | 444 | 67 | ||
XM_005550783.2 | XP_005550840.1 | 4 - 5 | 440 | 67 | ||
XM_015448047.1 | XP_015303533.1 | 5 - 6 | 489 | 67 | ||
GenBank mRNA | AB070189.1 | - | 4 - 5 | 588 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
