Assay Details
| Gene Symbol: ANKRD37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005556419.1 | XP_005556476.1 | 2 - 3 | 396 | 71 | |
| XM_005556420.2 | XP_005556477.1 | 2 - 3 | 412 | 71 | ||
More Information
Set Membership: |
|

ANKRD37
Genomic Map