Assay Details
| Gene Symbol: LOC102138027 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015452392.1 | XP_015307878.1 | 6 - 7 | 416 | 94 | |
| XM_015452393.1 | XP_015307879.1 | 6 - 7 | 540 | 94 | ||
More Information
Set Membership: |
|

LOC102138027
Genomic Map