Assay Details
| Gene Symbol: LOC102142032 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015456085.1 | XP_015311571.1 | 6 - 7 | 2236 | 93 | |
More Information
Set Membership: | 
		   
					
						
							 | 
				
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102142032| Gene Symbol: LOC102142032 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015456085.1 | XP_015311571.1 | 6 - 7 | 2236 | 93 | |
Set Membership: | 
		   
					
						
							 |