Assay Details
Gene Symbol: CCDC188 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005567937.2 | XP_005567994.1 | 5 - 6 | 1105 | 59 | |
XM_005567939.2 | XP_005567996.1 | 5 - 6 | 1103 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
