Assay Details
| Gene Symbol: LOC102117686 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005568171.2 | XP_005568228.1 | 2 - 3 | 470 | 91 | |
| XM_005568172.2 | XP_005568229.1 | 2 - 3 | 611 | 91 | ||
| XM_005568173.2 | XP_005568230.1 | 2 - 3 | 589 | 91 | ||
| XM_005568174.2 | XP_005568231.1 | 1 - 2 | 382 | 91 | ||
More Information
Set Membership: |
|

LOC102117686
Genomic Map