Assay Details
| Gene Symbol: LOC102134112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005583495.2 | XP_005583552.1 | 1 - 2 | 664 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102134112| Gene Symbol: LOC102134112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005583495.2 | XP_005583552.1 | 1 - 2 | 664 | 85 | |
Set Membership: |
|