Assay Details
| Gene Symbol: LOC102139474 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005590049.1 | XP_005590106.1 | 1 - 2 | 82 | 76 | |
| XM_005590050.1 | XP_005590107.1 | 1 - 2 | 82 | 76 | ||
| XM_005590051.1 | XP_005590108.1 | 1 - 2 | 82 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005590050.1 transcripts.
Set Membership: |
|

LOC102139474
Genomic Map