Assay Details
| Gene Symbol: CMTM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005592158.1 | XP_005592215.1 | 3 - 4 | 519 | 55 | |
| XM_015442889.1 | XP_015298375.1 | 3 - 4 | 432 | 55 | ||
More Information
Set Membership: |
|

CMTM4
Genomic Map