Assay Details
| Gene Symbol: MAGEB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005593224.1 | XP_005593281.1 | 1 - 2 | 69 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005593224.1 transcripts.
Set Membership: |
|

MAGEB2
Genomic Map