Assay Details
| Gene Symbol: ORAI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005549308.2 | XP_005549365.1 | 3 - 4 | 487 | 109 | |
| XM_005549309.2 | XP_005549366.1 | 3 - 4 | 494 | 109 | ||
More Information
Set Membership: |
|

ORAI2
Genomic Map