Assay Details
Gene Symbol: CRPPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005550062.2 | XP_005550119.1 | 1 - 2 | 434 | 73 | |
XM_005550063.2 | XP_005550120.1 | 1 - 2 | 434 | 73 | ||
XM_005550065.2 | XP_005550122.1 | 1 - 2 | 434 | 73 | ||
XM_015447681.1 | XP_015303167.1 | 1 - 2 | 434 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
