Assay Details
| Gene Symbol: ZFP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005558731.2 | XP_005558788.1 | 4 - 5 | 370 | 73 | |
| XM_005558733.2 | XP_005558790.1 | 4 - 5 | 654 | 73 | ||
| XM_005558734.2 | XP_005558791.1 | 3 - 4 | 527 | 73 | ||
| XM_005558735.2 | XP_005558792.1 | 2 - 3 | 494 | 73 | ||
| XM_015452355.1 | XP_015307841.1 | 3 - 4 | 633 | 73 | ||
| XM_015452356.1 | XP_015307842.1 | 3 - 4 | 630 | 73 | ||
| XM_015452357.1 | XP_015307843.1 | 2 - 3 | 888 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005558735.2, XM_015452357.1 transcripts.
Set Membership: |
|

ZFP2
Genomic Map