Assay Details
| Gene Symbol: LOC102118628 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005560872.2 | XP_005560929.1 | 11 - 12 | 1164 | 109 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102118628| Gene Symbol: LOC102118628 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005560872.2 | XP_005560929.1 | 11 - 12 | 1164 | 109 | |
Set Membership: |
|