Assay Details
| Gene Symbol: PRORP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005561069.2 | XP_005561126.1 | 5 - 6 | 1891 | 85 | |
| XM_005561070.2 | XP_005561127.1 | 5 - 6 | 1680 | 85 | ||
| XM_005561071.2 | XP_005561128.1 | 5 - 6 | 1673 | 85 | ||
| XM_005561072.2 | XP_005561129.1 | 4 - 5 | 1843 | 85 | ||
| XM_015453707.1 | XP_015309193.1 | 5 - 6 | 1677 | 85 | ||
| XM_015453708.1 | XP_015309194.1 | 2 - 3 | 252 | 85 | ||
| GenBank mRNA | AB179401.1 | - | 5 - 6 | 1896 | 85 | |

PRORP
Genomic Map