Assay Details
| Gene Symbol: CCN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005569089.2 | XP_005569146.1 | 4 - 5 | 2302 | 79 | |
| XM_015429869.1 | XP_015285355.1 | 3 - 4 | 2462 | 79 | ||
More Information
Set Membership: |
|

CCN5
Genomic Map