Assay Details
| Gene Symbol: APOA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005579727.2 | XP_005579784.1 | 1 - 2 | 216 | 97 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005579727.2 transcripts.
Set Membership: |
|

APOA4
Genomic Map