Assay Details
| Gene Symbol: ACRV1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005580084.2 | XP_005580141.1 | 3 - 4 | 1075 | 66 | |
| XM_005580086.2 | XP_005580143.1 | 3 - 4 | 973 | 66 | ||
More Information
Set Membership: |
|

ACRV1
Genomic Map