Assay Details
| Gene Symbol: PRRT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005591580.2 | XP_005591637.2 | 2 - 3 | 1183 | 69 | |
| XM_015442416.1 | XP_015297902.1 | 2 - 3 | 1122 | 69 | ||
| XM_015442417.1 | XP_015297903.1 | 2 - 3 | 1183 | 69 | ||
More Information
Set Membership: |
|

PRRT2
Genomic Map