Assay Details
| Gene Symbol: LOC102132457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015443982.1 | XP_015299468.1 | 1 - 2 | 99 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102132457| Gene Symbol: LOC102132457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015443982.1 | XP_015299468.1 | 1 - 2 | 99 | 64 | |
Set Membership: |
|