Assay Details
| Gene Symbol: PCDHB16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005595989.2 | XP_005596046.2 | 1 | 3420 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PCDHB16| Gene Symbol: PCDHB16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005595989.2 | XP_005596046.2 | 1 | 3420 | 70 | |
Set Membership: |
|