Assay Details
| Gene Symbol: SLC66A1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005546210.2 | XP_005546267.2 | 3 - 4 | 303 | 124 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC66A1L| Gene Symbol: SLC66A1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005546210.2 | XP_005546267.2 | 3 - 4 | 303 | 124 | |
Set Membership: |
|