Assay Details
| Gene Symbol: LOC102132357 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005547285.1 | XP_005547342.1 | 3 - 4 | 389 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102132357| Gene Symbol: LOC102132357 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005547285.1 | XP_005547342.1 | 3 - 4 | 389 | 58 | |
Set Membership: |
|