Assay Details
| Gene Symbol: LOC102124488 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005559573.2 | XP_005559630.1 | 3 - 4 | 756 | 132 | |
| XM_015452834.1 | XP_015308320.1 | 3 - 4 | 756 | 132 | ||
More Information
Set Membership: |
|

LOC102124488
Genomic Map