Assay Details
| Gene Symbol: HDHD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005586761.2 | XP_005586818.1 | 4 - 5 | 624 | 58 | |
| XM_015439916.1 | XP_015295402.1 | 4 - 5 | 624 | 58 | ||
More Information
Set Membership: |
|

HDHD2
Genomic Map