Assay Details
| Gene Symbol: GPRC5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319434.1 | NP_001306363.1 | 2 - 3 | 1292 | 89 | |
| XM_015442462.1 | XP_015297948.1 | 2 - 3 | 1292 | 89 | ||
| GenBank mRNA | AB172418.1 | - | 2 - 3 | 1292 | 89 | |
More Information
Set Membership: |
|

GPRC5B
Genomic Map