Assay Details
| Gene Symbol: TOR1AIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005540130.2 | XP_005540187.1 | 5 - 6 | 1372 | 72 | |
| XM_005540131.2 | XP_005540188.1 | 5 - 6 | 1371 | 72 | ||
| XM_005540132.2 | XP_005540189.1 | 5 - 6 | 1369 | 72 | ||
| XM_005540133.2 | XP_005540190.1 | 4 - 5 | 1326 | 72 | ||
| XM_005540134.2 | XP_005540191.1 | 5 - 6 | 1373 | 72 | ||
| XM_005540135.2 | XP_005540192.1 | 4 - 5 | 1325 | 72 | ||
| XM_005540136.2 | XP_005540193.1 | 4 - 5 | 1323 | 72 | ||
| XM_005540137.2 | XP_005540194.1 | 5 - 6 | 1372 | 72 | ||
| XM_005540138.2 | XP_005540195.1 | 4 - 5 | 1326 | 72 | ||
| XM_015446383.1 | XP_015301869.1 | 4 - 5 | 1327 | 72 | ||
| GenBank mRNA | AB170489.1 | - | 4 - 5 | 864 | 72 | |
More Information
Set Membership: |
|

TOR1AIP1
Genomic Map