Assay Details
| Gene Symbol: ETV3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005541370.2 | XP_005541427.1 | 2 - 3 | 335 | 68 | |
| XM_005541371.2 | XP_005541428.1 | 2 - 3 | 321 | 68 | ||
| XM_015455337.1 | XP_015310823.1 | 2 - 3 | 335 | 68 | ||
| GenBank mRNA | AB172595.1 | - | 2 - 3 | 83 | 68 | |
More Information
Set Membership: |
|

ETV3
Genomic Map