Assay Details
| Gene Symbol: ITLN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015452773.1 | XP_015308259.1 | 7 - 8 | 973 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ITLN2| Gene Symbol: ITLN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015452773.1 | XP_015308259.1 | 7 - 8 | 973 | 85 | |
Set Membership: |
|