Assay Details
| Gene Symbol: FILIP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005548313.2 | XP_005548370.1 | 3 - 4 | 889 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FILIP1L| Gene Symbol: FILIP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005548313.2 | XP_005548370.1 | 3 - 4 | 889 | 84 | |
Set Membership: |
|