Assay Details
| Gene Symbol: BRI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005549107.2 | XP_005549164.1 | 1 - 2 | 154 | 103 | |
| XM_005549108.2 | XP_005549165.1 | 1 - 2 | 154 | 103 | ||
| XM_005549109.2 | XP_005549166.1 | 1 - 2 | 154 | 103 | ||
More Information
Set Membership: |
|

BRI3
Genomic Map