Assay Details
| Gene Symbol: POP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005549251.2 | XP_005549308.1 | 2 - 3 | 338 | 94 | |
| XM_005549252.2 | XP_005549309.1 | 2 - 3 | 315 | 94 | ||
| XM_015447168.1 | XP_015302654.1 | 2 - 3 | 121 | 94 | ||
| XM_015447169.1 | XP_015302655.1 | 2 - 3 | 399 | 94 | ||
| XM_015447170.1 | XP_015302656.1 | 2 - 3 | 123 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005549251.2, XM_005549252.2, XM_015447168.1, XM_015447169.1, XM_015447170.1 transcripts.
Set Membership: |
|

POP7
Genomic Map