Assay Details
| Gene Symbol: LOC102124330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005555080.2 | XP_005555137.1 | 3 - 4 | 330 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102124330| Gene Symbol: LOC102124330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005555080.2 | XP_005555137.1 | 3 - 4 | 330 | 98 | |
Set Membership: |
|