Assay Details
| Gene Symbol: SLCO1B7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015430187.1 | XP_015285673.1 | 13 - 14 | 1705 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLCO1B7| Gene Symbol: SLCO1B7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015430187.1 | XP_015285673.1 | 13 - 14 | 1705 | 110 | |
Set Membership: |
|