Assay Details
Gene Symbol: FKBP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005570730.2 | XP_005570787.1 | 5 - 6 | 1228 | 66 | |
XM_015430865.1 | XP_015286351.1 | 5 - 6 | 1325 | 66 | ||
XM_015430866.1 | XP_015286352.1 | 4 - 5 | 428 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
