Assay Details
| Gene Symbol: KRT78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005570948.2 | XP_005571005.1 | 3 - 4 | 708 | 70 | |
| XM_015430962.1 | XP_015286448.1 | 3 - 4 | 332 | 70 | ||
More Information
Set Membership: |
|

KRT78
Genomic Map